Learn More
MilliporeSigma Rink Amide MBHA resin (100-200 mesh), 25G, 431041-83-7, MFCD00165781
Supplier:  MilliporeSigma 8550030025
Encompass
This support comprises the modified Rink amide linker attached via norleucine to MBHA resin, and is an ideal tool for the Fmoc SPPS of peptide amides. Cleavage from this resin can be effected by a single step treatment with 95% TFA, providing peptide amides in high yields and purities. Alternate Names: 4-(2',4'-Dimethoxyphenyl-Fmoc-aminomethyl)-phenoxyacetamido-norleucyl-MBHA resin (100-200 mesh).
Catalog No. NC1047059