Learn More
ABclonal Technology RIOK2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A12122500UL
Encompass_Preferred
Predicted to enable protein kinase activity. Involved in several processes, including positive regulation of rRNA processing, positive regulation of ribosomal small subunit export from nucleus, and regulation of mitotic metaphase/anaphase transition. Located in cytoplasm. Part of preribosome, small subunit precursor.
Catalog No. 50-001-14651