Learn More
Medchemexpress LLC RIP1 kinase inhibitor 1 | 2095515-38-9 | 99.96% | 461.90 | 10 MG
SDP
Supplier:  Medchemexpress LLC 10MGHY111409
Encompass_Preferred
RIP1 kinase inhibitor 1 (compound 22) is a highly potent, orally available, and brain-penetrating RIP1 kinase inhibitor with a pKi of 9.04. It strongly suppresses necroptotic cell death and phosphorylation of MLKL in human colorectal adenocarcinoma HT-29 cells and mouse L-cells NCTC 929.
  • Highly potent and orally available
  • Brain-penetrating RIP1 kinase inhibitor
  • Suppresses necroptotic cell death
  • Inhibits phosphorylation of MLKL
  • Effective in human colorectal adenocarcinoma HT-29 cells
  • Effective in mouse L-cells NCTC 929
Catalog No. 50-003-65055