Learn More
Medchemexpress LLC RIP2 kinase inhibitor 2 | 1581270-11-2 | 99.7% | 432.54 | 25 MG

Supplier: Medchemexpress LLC 25MGHY19761
RIP2 kinase inhibitor 2 is a receptor interacting protein-2 (RIP2) kinase inhibitor extracted from patent WO/2014043437 A1, compound example 9. It is a novel prodrug of a quinazolyl amine that inhibits RIP2 kinase. Receptor interacting protein-2 (RIP2) kinase is a TKL family serine/threonine protein kinase involved in innate immune signaling. Following activation, RIP2 kinase associates with NODI or NOD2 and appears to function principally as a molecular scaffold to bring together other kinases (TAK1, IKKα/β/γ) involved in NF-κB and mitogen-activated protein kinase activation. This product is for research use only and is not sold to patients.
- Target: RIP2 kinase
- Mechanism: Inhibits RIP2 kinase
- Source: Extracted from patent WO/2014043437 A1
- Molecular weight: 432.54
- Formula: C21H28N4O4S
- Cas no.: 1581270-11-2
- Appearance: Solid
- Color: Light yellow to yellow
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.