Learn More
Medchemexpress LLC RIP2 Kinase Inhibitor 3 | 1398053-50-3 | 99.8% | 388.48 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY112907100MG
Encompass_Preferred
RIP2 Kinase Inhibitor 3 is a highly potent and selective inhibitor of receptor interacting protein-2 (RIP2) Kinase with an IC50 of 1 nM. This product is for research use only and not intended for sale to patients.
  • Highly potent and selective inhibitor of RIP2 kinase
  • IC50 of 1 nM (RIP2)
Catalog No. 50-004-39812