Learn More
Abcam RIPA Buffer, 100ML
Supplier:  Abcam AB288006100ML
Encompass_Preferred
A ready-to-use buffer useful for lysing cells and tissues for radioimmunoprecipitation assay.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-3947