Learn More
Selleck Chemical LLC Ripasudil (K-115) hydrochloride dihydrate 5mg 887375-67-9
SDP
Supplier:  Selleck Chemical LLC S79955MG
Encompass_Preferred
Ripasudil (K-115) hydrochloride dihydrate is potent ROCK inhibitor with IC50 of 51 nM and 19 nM for ROCK1 and ROCK2, respectively, used for the treatment of glaucoma and ocular hypertension. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6407