Learn More
Medchemexpress LLC Ripgbm | 355406-76-7 | 100.0% | 25 MG
SDP
Supplier:  Medchemexpress LLC HY12291025MG
Encompass_Preferred
RIPGBM is a small-molecule research compound that selectively induces apoptosis in glioblastoma multiforme (GBM) cancer stem cells, with an EC50 ≤500 nM for induction of apoptosis. For research use only; not for human use.
  • Selective inducer of apoptosis in GBM cancer stem cells
  • Reported EC50 ≤500 nM for apoptosis in GBM CSCs
  • High purity suitable for research applications
  • Available as solids (5-500 mg) and 10 mM DMSO solutions
  • Supplied with data sheet, certificate of analysis, and safety data sheet
Catalog No. 50-002-30332