Learn More
Medchemexpress LLC Ripgbm | 355406-76-7 | 100.0% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY12291050MG
Encompass_Preferred
RIPGBM is a small-molecule apoptosis inducer that selectively triggers apoptosis in glioblastoma multiforme (GBM) cancer stem cells, with a reported EC50 of ≤500 nM. It is supplied as a solid for laboratory research use.
  • Selective apoptosis induction in GBM cancer stem cells (EC50 ≤500 nM).
  • Molecular formula C26H21FN2O3; molecular weight 428.45.
  • High reported purity (≈99.95%).
  • Available in multiple pack sizes, including 50 mg solid packs.
  • Intended for research use only; not for human or veterinary use.
Catalog No. 50-002-31326