Learn More
TARGETMOL CHEMICALS INC RIPK1-IN-9 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T630555MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. RIPK1-IN-9 is a potent and selective inhibitor of RIPK1 a dihydronaphthone compound which inhibits U937 and L929 cells with an IC50 of 2 nM and 1.3 nM respectively. purity: 98%
Catalog No. 50-257-9444