Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Recombinant human RIPK2 protein (P. pastoris, His) | >90.0% | 100UG
SDP

Supplier:  Medchemexpress LLC HYP700624100UG

Encompass_Preferred

Recombinant full-length human RIPK2 is supplied as a C-terminal His-tagged, lyophilized protein expressed in Pichia pastoris for research applications. The preparation is provided in a stabilizing buffer and is suitable for biochemical characterization and binding studies; enzymatic activity testing is in progress.

  • Full-length human RIPK2, residues M1-M540.
  • Expressed in Pichia pastoris and purified with a C-terminal 6xHis tag.
  • Purity >90% by reducing SDS-PAGE.
  • Approximate molecular weight 66.0 kDa.
  • Lyophilized from 10 mM Tris-HCl, 1 mM EDTA, 6% trehalose, pH 8.0.
  • Available in multiple package sizes (20, 50, 100 μg); larger quantities by quote.
  • Store at -20°C; aliquot and freeze at -20°C or -80°C for extended storage.
  • For research use only; enzyme activity testing in progress.

Catalog No. 50-001-76141


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.