Learn More
Aviva Systems Biology Corp RIPK4 Antibody - N-terminal region (ARP45871_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45871_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RIPK4. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human RIPK4. Alternate names: ANKK2, ANKRD3, DIK, MGC129992, MGC129993, PKK, RIP4, NKRD3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-221