Learn More
TARGETMOL CHEMICALS INC RIPRETINIB 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T84825MG
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ripretinib (DCC-2618) is an orally bioavailable inhibitor of KIT and PDGFRA. purity: 99%
Catalog No. 50-257-9068