Learn More
TARGETMOL CHEMICALS INC RISDIPLAM 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T167575MG
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Risdiplam (RO7034067) (RG7916) is orally administered. Risdiplam is a centrally and peripherally distributed SMN2 pre-mRNA splicing modifier which increases survival motor neuron (SMN) protein levels[1]. purity: 99%
Catalog No. 50-257-7328