Learn More
Apexbio Technology LLC Risperidone(Synonyms: Risperdal, Risperdal Consta, Belivon, Rispen, Risperin, Rispolept, Ridal, Risnia, Risperdone, Risperdaloro, Sequinan), 100mg, CAS: 106266-06-2.
SDP
Supplier:  Apexbio Technology LLC A8514100
Encompass_Preferred
Risperidone (CAS 106266-06-2) is an atypical antipsychotic small molecule that functions predominantly as an antagonist at serotonin receptor subtype 5-HT2A and dopamine receptor D2 (D2DR) It exhibits high receptor binding affinity with inhibitory constants (Ki) of approximately 0 4 nM for 5-HT2A receptors and 3 13 nM for D2 receptors Due to these pharmacological properties risperidone serves as a valuable research tool to study neurotransmitter modulation and receptor signaling pathways associated with psychiatric disorders particularly schizophrenia and related psychoses
Catalog No. 50-000-60069