Learn More
TARGETMOL CHEMICALS INC Risperidone 100MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T0351100MG
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Risperidone (R 64 766) is a selective blocker of dopamine D2 receptors and serotonin 5-HT2 receptors that act as an atypical antipsychotic agent. Purity 100%
Catalog No. 50-288-0518