Learn More
Medchemexpress LLC Risperidone | 106266-06-2 | 98% | 410.48 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11018100MG
Encompass_Preferred
Risperidone (Standard) is the analytical standard of Risperidone, intended for research and analytical applications. It functions as a serotonin 5-HT2 receptor blocker, a P-Glycoprotein inhibitor, and a potent dopamine D2 receptor antagonist, with Kis values of 4.8 nM for 5-HT2A and 5.9 nM for dopamine D2 receptor. It is for research use only and not sold to patients.
  • Analytical standard grade, supplied for assay reference
  • Commonly used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS
Catalog No. 50-003-44239