Learn More
Medchemexpress LLC Risperidone | 106266-06-2 | 99.9% | 410.48 | 10 MG
SDP
Supplier:  Medchemexpress LLC 10MGHY11018
Encompass_Preferred
Risperidone is a serotonin 5-HT2 receptor blocker, P-Glycoprotein inhibitor, and potent dopamine D2 receptor antagonist, with Ki values of 4.8 nM for 5-HT2A and 5.9 nM for dopamine D2 receptor. It is intended for research use only.
  • Dose-dependently inhibits the release of IL-12 in mature DCs.
  • Dose-dependently increases the production of IL-10.
  • High doses can induce TNF-α release from mature DCs.
  • Causes slightly but significantly lower body weight in rats as a function of age.
Catalog No. 50-003-65037