Learn More
Medchemexpress LLC Risperidone | 106266-06-2 | MFCD00274576 | 99.9% | 410.48 g·mol⁻¹ | C23H27FN4O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY110185MG
Encompass_Preferred
Risperidone is an antipsychotic research compound used in laboratory and analytical applications. It functions as a serotonin 5-HT2A receptor blocker, a P-glycoprotein inhibitor, and a potent dopamine D2 receptor antagonist, and is provided as a high-purity reference material for in vitro studies.
  • Acts as a 5-HT2A receptor blocker and dopamine D2 antagonist.
  • Demonstrated high binding affinity (Ki ≈ 4.8-5.9 nM for 5-HT2A and D2).
  • Supplied in small research quantities suitable for assay development.
  • High chemical purity appropriate for analytical work (purity 99.87% by HPLC).
  • Molecular formula C23H27FN4O2; molecular weight 410.48 g·mol⁻¹.
Catalog No. 50-002-63778