Learn More
Medchemexpress LLC Risperidone | 106266-06-2 | MFCD00274576 | 99.9% | 410.5 g/mol | 100 MG
SDP
Supplier:  Medchemexpress LLC HY1101850MG
Encompass_Preferred
Risperidone is a serotonin 5-HT2 receptor blocker, P-Glycoprotein inhibitor and potent dopamine D2 receptor antagonist, with Kis of 4.8, 5.9 nM for 5-HT2A and dopamine D2 receptor, respectively. It is for research use only.
  • Serotonin 5-HT2 receptor blocker
  • P-Glycoprotein inhibitor
  • Potent dopamine D2 receptor antagonist
  • For research use only
Catalog No. 50-003-44151