Learn More
Cayman Chemical RIsutegnIbtrIfluoroacetAT 10mg
SDP
Supplier:  Cayman Chemical 3330310 MG
Encompass_Preferred
An anti-integrin peptide; protects against hydroquinone-induced necrosis and apoptosis, as well as inhibits hydroquinone-induced production of ROS in RPE cells at 400 µM; decreases expression of BAX, VEGFA, and ITB1 in human AMD cybrid cell lines at 20 mg/ml; arrests aberrant blood vessel growth mediated by αVβ3, αVβ5, and α5β1 integrins in vivo
Catalog No. 50-294-6988