Learn More
Selleck Chemical LLC RITA (NSC 652287) 5mg 213261-59-7
SDP
Supplier:  Selleck Chemical LLC S27815MG
Encompass_Preferred
RITA (NSC 652287) induces both DNA-protein and DNA-DNA cross-links with no detectable DNA single-strand breaks, and also inhibits MDM2-p53 interaction by targeting p53. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2951