Learn More
Medchemexpress LLC HY-100754 50mg , Ritlecitinib CAS:1792180-81-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10075450MG
Encompass_Preferred
HY-100754 50mg Ritlecitinib CAS: 1792180-81-4 Ritlecitinib (PF-06651600) is an orally active and selective JAK3 inhibitor with an IC50 of 33.1 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0380