Learn More
Medchemexpress LLC Ritlecitinib tosylate | 2192215-81-7 | MFCD25976759 | 99.8% | 457.55 g·mol⁻1 | C22H27N5O4S | 1 ML
SDP
Supplier:  Medchemexpress LLC HY100754C10MM 1ML
Encompass_Preferred
Ritlecitinib tosylate is the tosylate salt form of a selective JAK3 inhibitor supplied as high-purity research material. Offered in both solid and DMSO solution formats, including ready-to-use 10 mM solutions, it is intended for in vitro biochemical and cellular assays targeting JAK3-related pathways.
  • High purity suitable for research assays.
  • Available as solid and DMSO solution formats.
  • Ready-to-use 10 mM solution for rapid assay setup.
  • Supplied with datasheet, COA, and SDS for quality and safety.
  • Supports biochemical and cellular JAK3 research applications.
Catalog No. 50-002-11363