Learn More
Apexbio Technology LLC Ritodrine HCl 23239-51-2 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B13475.1
Encompass_Preferred
Ritodrine hydrochloride (CAS 23239-51-2) is a selective 2-adrenergic receptor agonist that modulates smooth muscle activity By stimulating 2-adrenergic receptors ritodrine promotes uterine muscle relaxation thereby inhibiting premature uterine contractions Comparative studies indicate that ritodrine can rapidly extend the interval before delivery although it is associated with a higher incidence of adverse effects compared to alternatives such as magnesium sulfate Ritodrine HCl is widely used in research to investigate 2-adrenergic signaling pathways and to model the pharmacological regulation of preterm labor
Catalog No. 50-000-88711