Learn More
Medchemexpress LLC Ritodrine hydrochloride | 23239-51-2 | 98.5% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB045210MM/1ML
Encompass_Preferred
Ritodrine hydrochloride is a potent and orally active β-adrenergic agonist. It is known to decrease preterm delivery and uterine contraction, making it a valuable compound for research into preterm labor.
  • Potent and orally active beta-adrenergic agonist
  • Decreases preterm delivery
  • Decreases uterine contraction
  • Potential for research of preterm labor
  • Targets beta adrenergic receptor
Catalog No. 50-004-05393