Learn More
Medchemexpress LLC Rivaroxaban | 366789-02-8 | MFCD11974010 | 99.5% (assay) / 99.7% (HPLC) | 435.88 | C19H18ClN3O5S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY50903R10MG
Encompass_Preferred
Rivaroxaban (Standard) is an analytical reference standard of the oral anticoagulant rivaroxaban (BAY 59-7939), supplied for research and analytical applications including method development, calibration, and quality control for HPLC and LC-MS assays.
  • Analytical reference standard for method development and validation.
  • High purity suitable for calibration and quality control.
  • Supplied as a solid in an amber glass vial to protect stability.
  • Characterized by CAS 366789-02-8 and formula C19H18ClN3O5S.
Catalog No. 50-002-98640