Learn More
Medchemexpress LLC Rivaroxaban impurity 68 | 845729-41-1 | 99.9% | C10H10N2O4 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY75562500MG
Encompass_Preferred
Rivaroxaban impurity 68 is a chemical compound with the formula C10H10N2O4, used as a reference standard in research and quality control applications. It has a purity of 99.89% and a CAS number of 845729-41-1. This high-purity impurity is essential for identifying and quantifying impurities in Rivaroxaban formulations.
  • High purity for accurate analytical results
  • Essential for pharmaceutical quality control
  • Ideal for research and development
  • Reliable reference standard
Catalog No. 50-004-33608