Learn More
Medchemexpress LLC Rivastigmine (Standard) | 123441-03-2 | 99.9% | 250.34 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY17368R50MG
Encompass_Preferred
Rivastigmine (Standard) is the analytical standard of Rivastigmine, intended for research and analytical applications. It is an orally active and potent cholinesterase (ChE) inhibitor. This compound is an analytical standard used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Orally active and potent cholinesterase (ChE) inhibitor
  • Inhibits butyrylcholinesterase (BChE) and acetylcholinesterase (AChE)
  • Can pass the blood-brain barrier (BBB)
  • Used for research of mild to moderate dementia of the Alzheimer's type and dementia due to Parkinson's disease
  • Analytical standard for HPLC, GC, and MS research experiments
  • Appearance: Liquid, colorless to light yellow
Catalog No. 50-003-84759