Learn More
Selleck Chemical LLC Rizatriptan Benzoate 10mg 145202-66-0 MK-462 Benzoate
SDP
Supplier:  Selleck Chemical LLC S160710MG
Encompass_Preferred
Rizatriptan Benzoate (MK-462) is an agonist at serotonin 5-HT1B and 5-HT1D receptors, used to treat acute migraine attacks. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-538-6