Learn More
Medchemexpress LLC Rjw100 | 1276664-20-0 | 99.9% | 386.58 | C28H34O | 5 MG

Supplier: Medchemexpress LLC HY1314455MG
RJW100 is a small-molecule agonist of liver receptor homolog-1 (LRH-1) and steroidogenic factor-1 (SF-1). It is supplied as a high-purity research reagent for biochemical and cell-based studies of nuclear receptor signaling, endocrinology, and cancer-related pathways.
- Potent LRH-1 and SF-1 agonist (pEC50 ≈6.6 and 7.5)
- High purity, approximately 99.9%
- Molecular formula C28H34O, molecular weight 386.58
- Supplied in small milligram quantities for research use
- Storage recommendations: keep cold to preserve stability
- For research use only, not for human or veterinary applications
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.