Learn More
Selleck Chemical LLC RK 24466 5mg
SDP
Supplier:  Selleck Chemical LLC S00205MG
Encompass_Preferred
RK-24466 (KIN 001-51, Lck inhibitor C 8863, C8863) is a potent and selective inhibitor of Lck that inhibits two constructs of the human lck kinase, lck (64-509) and lckcd with IC50 of < 0.001 ?M and 0.002 ?M, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3020