Learn More
Medchemexpress LLC RK-701 | 2648855-18-7 | C26H30N4O3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY152775100MG
Encompass_Preferred
RK-701 is a highly selective and non-genotoxic inhibitor of G9a, with an IC50 value of 23-27 nM. It selectively up-regulates HbF, γ-Globin, and BGLT3 expression, while down-regulating H3K9me2 expression. RK-701 also inhibits BCL11A and ZBTB7A, and is intended for research use only.
  • Highly selective G9a inhibitor
  • Non-genotoxic compound
  • Potent inhibition with IC50 value of 23-27 nM for G9a
  • Up-regulates HbF, γ-Globin, and BGLT3 expression
  • Down-regulates H3K9me2 expression
  • Inhibits BCL11A and ZBTB7A
  • Low toxicity observed in in vivo studies
Catalog No. 50-003-73668