Learn More
ABclonal Technology LbCas12a Nuclease
SDP
Supplier:  ABclonal Technology RK205701000PMOL
Encompass_Preferred
Lb Cas12a Nuclease from Lachnospiraceae bacterium is a site-specific DNA endonuclease guided by a single 40-44 nucleotide guide RNA (crRNA). Targeting requires a crRNA complementary to the target site as well as a 5’ TTTV protospacer adjacent motif (PAM) on the DNA strand opposite the target sequence. Cleavage by LbCas12a Nuclease occurs ~17 bases 3′ of the PAM and leaves overhanging ends.This product also has a trans-cleavage activity, which produces high non-specific ssDNA cleavage activity when the target dsDNA, crRNA, and Cas12a protein form a ternary complex.
Catalog No. 50-001-35804