Learn More
ABclonal Technology T4 DNA Ligase
SDP
Supplier:  ABclonal Technology RK2150180000U
Encompass_Preferred
T4 DNA Ligase catalyzes the formation of a phosphodiester bond between juxtaposed 5 phosphate and 3 hydroxyl termini in duplex DNA or RNA. This enzyme will join blunt end and cohesive end termini as well as repair single stranded nicks in duplex DNA and some DNA/RNA hybrids. T4 DNA ligase will seal nicks for these DNA substrates. T4 DNA Ligase is applicable to cloning of restriction fragments and to joining linkers and adapters to blunt-ended DNA.
Catalog No. 50-001-36638