Learn More
ABclonal Technology Fast Magnetic DNA Methylation Bisulfite Kit
SDP
Supplier:  ABclonal Technology RK3020650RXN
Encompass_Preferred
This kit quickly converts unmethylated cytosines in DNA samples to uracil while leaving methylated cytosines unchanged. After bisulfite treatment at high temperature, double-stranded DNA is denatured into single strands. Under the action of HSO3-, cytosine residues undergo deamination and are converted to uracil, while methylated residues remain intact. Subsequently, during PCR amplification, uracil is replaced by thymine (T).
Catalog No. 50-001-35042