Learn More
Medchemexpress LLC RKI-1447 dihydrochloride | 1782109-09-4 | 99.9% | 399.29 g/mol | C16H16Cl2N4O2S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1103395MG
Encompass_Preferred
RKI-1447 dihydrochloride is the dihydrochloride salt of a selective Rho-associated protein kinase (ROCK) inhibitor provided for laboratory research use. It is used in biochemical and cell-based assays to study ROCK signaling, cytoskeletal dynamics, and anti-tumor activity. The product is available as a solid and as a 10 mM solution in DMSO and is characterized by high purity.
  • High purity suitable for research assays.
  • Available as solid or 10 mM solution in DMSO.
  • Provided in small lab pack sizes for discovery research.
  • Store sealed at 4°C; in solution store at -80°C for long-term storage.
  • Intended for in vitro and cell-based research applications only.
Catalog No. 50-002-63525