Learn More
Abcam RKRARKE, PKG Inhibitor, 1MG
Supplier:  Abcam AB1450631MG
Encompass_Preferred
MW 943.12 g/mol, Purity >99%. Competitive protein kinase G (PKG) inhibitor. Ki = 86 µM for PKG; Ki = 550 µM for PKA. Potential inhibitory activity in neuronal systems.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5436