Learn More
Medchemexpress LLC Rl-6-me-7-oh | 2184-54-5 | 96.6% | 328.28 g/mol | C12H16N4O7 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15719410MG
Encompass_Preferred
RL-6-Me-7-OH is a small-molecule hapten that activates human and mouse mucosal-associated invariant T (MAIT) cells (reported EC50 = 25 μM). It is supplied as a solid research reagent for use in antigen design and biochemical assays, and is characterized by CAS RN 2184-54-5, molecular formula C12H16N4O7, and molecular weight 328.28 g/mol.
  • MAIT cell activator with reported EC50 = 25 μM.
  • Offered as a solid research reagent suitable for antigen design and biochemical assays.
  • Characterized by CAS RN 2184-54-5 and molecular formula C12H16N4O7.
  • Documentation available includes a datasheet, COA, and SDS for handling and characterization.
Catalog No. 50-002-87856