Learn More
Selleck Chemical LLC RMC-6291-E1606-1MG
SDP
Supplier:  Selleck Chemical LLC E16061MG
Encompass_Preferred
RMC-6291 is an orally active and covalent inhibitor of KRASG12C(ON) RMC-6291 forms a tri-complex within tumor cells between KRASG12C(ON) and cyclophilin A (CypA)
Catalog No. 50-313-7396