Learn More
TARGETMOL CHEMICALS INC RMC-9805 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T782121MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. RMC-9805 is a potent KRAS G12D inhibitor with potential anti-tumor and inhibitory activity against tumor cell proliferation for the study of pancreatic cancer. Purity 98.93%
Catalog No. 50-287-6232