Learn More
Cayman Chemical TOFA, 54857-86-2, 10 mg
SDP
Supplier:  Cayman Chemical 1000526310
Encompass_Preferred
Molecular Formula C19H32O4, Purity ≥98%, Formula Weight 324.5, Synonyms: RMI 14514, 5-(Tetradecyloxy)-2-furoic Acid; An inhibitor of fatty acid synthesis that blocks the synthesis of malonyl-CoA by acetyl-CoA carboxylase (ACC).
Catalog No. NC0291252