Learn More
ABclonal Technology RMND5A Rabbit pAb
SDP
Supplier:  ABclonal Technology A14924500UL
Encompass_Preferred
Predicted to enable metal ion binding activity and ubiquitin protein ligase activity. Predicted to contribute to ubiquitin-protein transferase activity. Predicted to be involved in proteasome-mediated ubiquitin-dependent protein catabolic process and protein polyubiquitination. Located in cytoplasm and nucleoplasm. Part of ubiquitin ligase complex.
Catalog No. 50-001-05219