Learn More
Mascon Medical RN Pipet Tips, 20 L, Stacked Refills, 960 tips in 10 layers of 96, Clear, Steile
SDP
Supplier:  Mascon Medical MM8600111P01
Encompass_Preferred
Compatibility:Rainin LTS SeriesFeaturesImported polypropylene(PP),USP Class VI standardsSmooth inner wall,reduce liquid residue,ensure the accuracy of liquid suctionDNA-free,DNase/RNase-free,Pyrogen/Endotoxin-freeUltra-grinding &molding technology,release agentfreeWithstand temperature ranging fom-80 to 121,stable perfomance underhigh temperature andhigh pressureExtrathin-wall design increase productflexibilityand improve heat transfer efficiency
Catalog No. NC3527218