Learn More
Apexbio Technology LLC RN486, 5mg. Cas: 1242156-23-5 MFCD: MFCD24682729. Btk inhibitor,potent and selective
SDP
Supplier:  Apexbio Technology LLC A86365
Encompass_Preferred
RN486 is a reversible inhibitor of Btk with IC50 value of 4.0 nM [1].Bruton's tyrosine kinase (Btk) is a type of kinase protein which is expressed by immune system-related cells and plays a pivotal role in cell differentiation and proliferation at the transition from pre-B to later B cell stages. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5172