Learn More
Zymo Research Corporation RNA Clean & Concentrator™-25 (100 Preps) w/ Zymo-Spin™ IIC Columns (Capped)
SDP
Supplier:  Zymo Research Corporation R1018
Encompass_Preferred
The RNA Clean & Concentrator-25 kits are RNA clean up kits that provide a simple and reliable method for the rapid preparation of high-quality, RT-PCR-ready RNA. This simple procedure is based on the use of a unique single-buffer system and Zymo-Spin column technology that allows for selective recovery of total RNA (> 17 nt), large RNAs (> 200 nt), and/or small RNAs (17-200 nt). The procedure is easy: Add binding buffer and ethanol to your sample, then bind, wash and elute ultra pure RNA. The RNA can be eluted from the Zymo-Spin IICR Column in as little as ≥ 25 µl of RNase-free water. The highly-concentrated, purified RNA is suitable for all subsequent analyses and molecular manipulations.
Catalog No. NC0938600