Learn More
Norgen Biotek Corp RNA/DNA/Protein Purification Plus Kit, 50 preps, spin-column method, processes in < 30 min, compatible with cultured animal cells, tissues, blood, yeast, bacteria, fungi, plants
Supplier:  Norgen Biotek Corp 47700
Encompass_Preferred
The Norgen Biotek Corp RNA/DNA/Protein Purification Plus Kit offers a rapid and efficient spin-column method for the isolation and purification of total RNA, genomic DNA, and proteins from a single biological sample. This kit is designed for researchers studying various biological topics while ensuring reliable analysis from precious or small samples. The purification process can be completed in less than 30 minutes, yielding high-integrity macromolecules suitable for a wide range of downstream applications such as real-time PCR, sequencing, and Western blots.
  • Sequential purification of total RNA, genomic DNA, and proteins from a single sample
  • Rapid processing completed in under 30 minutes
  • Compatible with numerous sample types, including cultured animal cells, blood, and plants
  • Produces high-purity RNA, DNA, and proteins ready for downstream applications
  • Comprehensive recovery of RNA sizes including microRNA
Catalog No. NC1953509