Learn More
Bioworld RNA Molecular Weight Marker, 18S + 28S Ribosomal RNA (2000-5300 bp), 200 μg
Supplier:  Bioworld 104300142
Encompass_Preferred
- Ultra Pure Grade - Ribosomal RNA, purified from a rat, consisting of 2 fragments approximately 2,000 bp and 5,300 bp in length. Function Test(P/F)- Pass RNase(P/F)- None
Catalog No. 50-196-434