Learn More
Medchemexpress LLC RNA Polymerase II Subunit B3 Antibody (YA2447) | 10 UL
SDP
Supplier:  Medchemexpress LLC HYP8270210UL
Encompass_Preferred
The RNA Polymerase II Subunit B3 Antibody (YA2447) is a rabbit-derived, non-conjugated IgG monoclonal antibody designed to target RNA Polymerase II Subunit B3. This antibody is suitable for applications such as Western Blot (WB) and Immunohistochemistry-Paraffin (IHC-P), making it a valuable tool for research in epigenetics and nuclear signaling. It targets the third largest subunit of RNA polymerase II, which is crucial for synthesizing messenger RNA in eukaryotes.
  • Targets RNA Polymerase II Subunit B3
  • Rabbit-derived IgG monoclonal antibody
  • Non-conjugated
  • Suitable for WB and IHC-P applications
  • Reactivity with human, mouse, and rat samples
  • Affinity purified
  • Stored at -20°C for 1 year
Catalog No. 50-004-43171