Learn More
Gold Biotechnology Inc RNase A
SDP
Supplier:  Gold Biotechnology Inc R0501
Encompass_Preferred
RNase A isolated from bovine pancreas degrades single-stranded RNA at the C and U residues therefore RNase A is pyrimidine specific Degradation occurs by cleaving the phosphodiester bond located between the 5 -ribose of the nucleotide and the phosphate group that is connected to the 3 -ribose of the neighboring pyrimidine
Catalog No. 50-005-62129